Order Entry
Switzerland
ContactUsLinkComponent
Toluene-α-sulphonyl fluoride 99%
Toxic
Corrosive
Toluene-α-sulphonyl fluoride 99%
Certificates
undefined
Toxic
Corrosive
Toluene-α-sulphonyl fluoride 99%
Certificates

Specification Test Results

  • Appearance:
    White Needles and/or agglomerates
  • Infrared spectrum:
    Conforms
  • Melting point:
    90 to 94 °C
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together