3,4-Dehydro-L-proline ≥99.0% (by HPLC)
TCIAD3825-100MG
: TCI
:
- Pk:100 mg
- Seq:2440
- Appearance:White to Amber to Dark green powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
- Specific rotation [a]20/D:-380.0 to -410.0 deg (C=1, H2O)
:
:113.12 g/mol
:223-738-7
:4043-88-3
:C₅H₇NO₂
:MFCD00065964