
3-(Dimethylamino)propanoic acid hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:14788-12-6
:
Formula:C5H12ClNO2
MDL Number:MFCD02093464
:153.6073 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:186.0 to 192.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together