
Sodium dodecylbenzenesulphonate (mixture) 62.00% wetted with water, soft type
Supplier: TCI
Certificates
About this item
CAS Number:69669-44-9
:
Formula:C₁₈H₃₀O₃S
MDL Number:MFCD00011508
:348.48 g/mol
Merck Index:14,08612
EINECS:274-070-8
:GHS07
Specification Test Results
- Concentration (Phase splitting method):62.0 to 68.0 w/w%
Specification Test Results
Frequently Bought Together