Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Methyl palmitate

Glass bottle for solids, 25 g
Catalog # ROTH4459.1
Marketsource
Supplier:   Roth Carl
Methyl palmitate
Catalog # ROTH4459.1
Supplier:  Roth Carl
Marketsource
CAS Number:  112-39-0
Methyl palmitate

Glass bottle for solids, 25 g

CAS Number:  112-39-0
Price
SFr. 36.60
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Hexadecylic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester, Hexadecanoic acid methyl ester, Palmitic Acid Methyl Ester
  • CAS Number:112-39-0
  • Formula:C₁₇H₃₄O₂
  • MDL Number:MFCD00008994
  • Molecular Weight:270.46 g/mol
  • Boiling Pt:417 °C (1013 hPa)
  • Melting Pt:29.9 °C
  • Density:0.8804 g/cm³ (20 °C)
  • EINECS:203-966-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications