Order Entry
Switzerland
ContactUsLinkComponent
Methyl palmitate for synthesis, Sigma-Aldrich®
Methyl palmitate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Methyl palmitate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) practically insoluble
  • Melting Point:
    30 °C
  • Molar Mass:
    270.46 g/mol
  • Boiling Point:
    415 - 418 °C (996 hPa)
  • Vapor Pressure:
     - 0.01 hPa (20 °C)
  • Flash Point:
    113 °C

Specification Test Results

Related Information

Frequently Bought Together