Methyl enantate
Supplier: MATREYA
Jump to:
About this item
Synonyms: Methyl Heptanoate, Methyl Enanthate, Enanthic acid methyl ester, Heptanoic acid methyll ester, Heptylic acid methyl ester, Heptanoic acid methyl ester, Enanthic acid methyll ester, Methyl heptanoate
- CAS Number:106-73-0
- Formula:C₈H₁₆O₂
- MDL Number:MFCD00009537
- Molecular Weight:144.21 g/mol
- Merck Index:12,04695
- Boiling Pt:172 °C (1013 hPa)
- Melting Pt:–56 °C
- Density:0.88 g/cm³ (20 °C)
- EINECS:203-428-8
1 Options Available Below
Product Details & Documents
Synonyms: Methyl Heptanoate, Methyl Enanthate, Enanthic acid methyl ester, Heptanoic acid methyll ester, Heptylic acid methyl ester, Heptanoic acid methyl ester, Enanthic acid methyll ester, Methyl heptanoate
- CAS Number:106-73-0
- Formula:C₈H₁₆O₂
- MDL Number:MFCD00009537
- Molecular Weight:144.21 g/mol
- Merck Index:12,04695
- Boiling Pt:172 °C (1013 hPa)
- Melting Pt:–56 °C
- Density:0.88 g/cm³ (20 °C)
- EINECS:203-428-8



