Order Entry
Switzerland
ContactUsLinkComponent
 
2-Ethylhexyl acetate for synthesis, Sigma-Aldrich®
 :  Merck
 
2-Ethylhexyl acetate for synthesis, Sigma-Aldrich®
 :  Merck
 

 

  • Solubility:
    (20 °C) practically insoluble
  • Melting Point:
    -92 °C
  • Molar Mass:
    172.26 g/mol
  • Boiling Point:
    199 °C (1013 hPa)
  • Vapor Pressure:
    0.28 hPa (20 °C)
  • Flash Point:
    85 °C
  • Refractive Index:
    1.4204 (20 °C, 589 nm)
  • Explosion Limit:
    0.76 - 8.14 %(V)
  • Density:
    0.872 g/cm3 (20 °C)
  • Ignition Point:
    268 °C

 

Frequently Bought Together