2-Ethylhexyl acetate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:103-09-3
:
Formula:C₁₀H₂₀O₂
MDL Number:MFCD00027249
:172.27 g/mol
Merck Index:14,6763
Boiling Pt:199…200 °C
Melting Pt:–45 °C
Density:0.870
Flash Pt:76 °C (169 °F)
Storage Temperature:Ambient
EINECS:203-079-1
Related Information
Frequently Bought Together