Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Methyl decanoate

Methyl decanoate

Supplier:   MATREYA
Methyl decanoate
Supplier:  MATREYA
Certificates
Methyl decanoate

1 Options Available Below

Product Details & Documents
Synonyms:  Decanoic acid methyl ester, Capric acid methyl ester, Decylic acid methyl ester, Methyl caprate
  • CAS Number:110-42-9
  • Formula:C₁₁H₂₂O₂
  • MDL Number:MFCD00009580
  • Molecular Weight:186.29 g/mol
  • Boiling Pt:224 °C (1013 hPa)
  • Melting Pt:–18 °C
  • Density:0,8724 g/cm³ (20 °C)
  • Flash Pt:94 °C
  • EINECS:203-766-6
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg

  • Glass bottle for solids
    500 mg
    Discontinued
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.