Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
Bis(4-hydroxy-3-methylphenyl)sulphide ≥98.0% (by GC)
  TCIAB1527-25G
 :  TCI
 :  
 
 
Bis(4-hydroxy-3-methylphenyl)sulphide ≥98.0% (by GC)
  TCIAB1527-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    122.0 to 125.0 deg-C
  • Solubility in Methanol:
    almost transparency

 

 

 :
 :246.32 g/mol
 :403-330-7
 :3077
 :9,III
 :GHS05
 :GHS09
 :24197-34-0
 :C₁₄H₁₄O₂S
 :MFCD00142117