Order Entry
Switzerland
ContactUsLinkComponent
Adenosine-3',5'-cyclic monophosphate sodium salt ≥98.0% (by HPLC)
Adenosine-3',5'-cyclic monophosphate sodium salt ≥98.0% (by HPLC)
Catalog # TCIAA2112-100MG
Supplier:  TCI
CAS Number:  
Adenosine-3',5'-cyclic monophosphate sodium salt ≥98.0% (by HPLC)
Catalog # TCIAA2112-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Water:
    max. 10.0 %

Specification Test Results

About this item

 :
 :351.19 g/mol
CAS Number:37839-81-9
Formula:C₁₀H₁₁N₅NaO₆P
MDL Number:MFCD00069736