Divinyl adipate ≥99.0% (by GC) stabilised
: TCI
:4074-90-2
:
:C₁₀H₁₄O₄
:MFCD00059202
:Stabilised with 4-methoxyphenol (MEHQ)
:198.22 g/mol
:Refrigerator
:223-792-1
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 99.0 %
- Solubility in Methanol:almost transparency
Frequently Bought Together