Order Entry
Switzerland
ContactUsLinkComponent
3-Chloro-2-methylaniline hydrochloride ≥98.0% (by titrimetric analysis)
Irritant
3-Chloro-2-methylaniline hydrochloride ≥98.0% (by titrimetric analysis)
Catalog # TCIAA0285-25G
Supplier:  TCI
CAS Number:  
Irritant
3-Chloro-2-methylaniline hydrochloride ≥98.0% (by titrimetric analysis)
Catalog # TCIAA0285-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (Volhard Method):
    min. 98.0 %
  • Solubility in Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :178.06 g/mol
EINECS:228-402-3
 :GHS07
CAS Number:6259-40-1
Formula:C₇H₈ClN
MDL Number:MFCD00058279