


1,4-Dioxane ≥99% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:123-91-1
:
Formula:C₄H₈O₂
MDL Number:MFCD00006571
:88.11 g/mol
Merck Index:14,3300
Boiling Pt:100…102 °C
Melting Pt:11.8 °C
Density:1.034
Flash Pt:12 °C (54 °F)
Storage Temperature:Ambient
UN:1165
ADR:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 5-10 ppm
EINECS:204-661-8
:GHS02
:GHS08
:GHS07
Related Information
Frequently Bought Together