ContactUsLinkComponent
Petrochemical distillation apparatus for water in crude oil
Petrochemical distillation apparatus for water in crude oil
Codice catalogo 201-1418
Nome del fornitore:  DWK Life Sciences
Petrochemical distillation apparatus for water in crude oil
Codice catalogo 201-1418
Nome del fornitore:  DWK Life Sciences
Specifiche
  • Description:
    Petrochemical distillation apparatus, complete
  • Pk:
    1
Specifiche
Informazioni su questo articolo
This apparatus is used for the determination of water in crude oil by distillation.
  • Designed from ASTM D4006 specifications
  • Manufactured from borosilicate 3.3 glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Apparatus includes a 1000 ml round bottom flask with NS 24/40 joint, a distillation receiver with 0,05 graduations, a drying trap and a 400 ml Liebig condenser.