Disodium hydrogen phosphate dihydrate
Catalog # ROTHT877.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
Specifications
About this item
:
:177.99 g/mol
Melting Pt:92.5 °C
Density:2.1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-448-7
CAS Number:10028-24-7
Formula:Na₂HPO₄·2H₂O
MDL Number:MFCD00149182