D-(-)-Serine
Marketsource
Supplier: BACHEM BIOCHEMICA
Jump to:
About this item
Synonyms: (R)-(-)-Serine, H-D-Ser-OH
- CAS Number:312-84-5
- Formula:C₃H₇NO₃
- MDL Number:MFCD00004269
- Molecular Weight:105.09 g/mol
- Merck Index:12,08604
- Melting Pt:235 °C
- EINECS:206-229-4
2 Options Available Below
Product Details & Documents
Synonyms: (R)-(-)-Serine, H-D-Ser-OH
- CAS Number:312-84-5
- Formula:C₃H₇NO₃
- MDL Number:MFCD00004269
- Molecular Weight:105.09 g/mol
- Merck Index:12,08604
- Melting Pt:235 °C
- EINECS:206-229-4
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



