Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
 
 
Hexachloroplatinic(IV) acid hydrate ACS
 
undefined
 
 
 
 
Hexachloroplatinic(IV) acid hydrate ACS

 

  • Appearance:
    Orange to orange-brown Powder and/or chunks
  • Assay:
    ≥37.50 % (Pt)
  • Solubility:
    (in alcohol) Passes test
  • Suitability:
    (for K determination) Passes test
  • Impurity:
    ≤0.05 % Alkali and other salts (as sulfates)
  • Acidity (CH3COOH):
    ≤0.001 %

 

 

Frequently Bought Together