Potassium permanganate 0.01 M KMnO4 (0.05N) volumetric solution, Fluka™
Catalog # 34266-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Factor:0.998 - 1.002
Specification Test Results
About this item
:
:158.03 g/mol
Boiling Pt:100 °C (1.013 hPa)
Melting Pt:0 °C
Density:1.0 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-760-3
ADR:9,III
:GHS09
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364