SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Sulphuric acid
Sulphuric acid 0.5 mol/l (1 N) in aqueous solution
Sulphuric acid 0.5 mol/l (1 N) in aqueous solution
:
Hach
Sulphuric acid 0.5 mol/l (1 N) in aqueous solution
:
Hach
:
7664-93-9
:
:
H₂SO₄
:
MFCD00064589
:
98.08 g/mol
:
1.01 g/cm³ (20 °C)
:
Ambient
:
231-639-5
:
2796
:
8,II
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
: Merck
Buffer solutions in sachets, Certipur®, Supelco®
: Merck
Dip slides for surfaces and in liquids, Envirocheck®
: Merck
Fluoride standard solution, 1,000 mg/L F- in Water (from NaF) Certipur®, Supelco®
: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
: Merck
VWR®, Primary colour solutions, Reagent Ph. Eur.
: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals
Starch 1% in aqueous solution stabilised Reag. Ph. Eur. 1085103
: VWR Chemicals
Powder pillows, reagents for Hach instruments
: Hach
Powder pillows, silica
: Hach