Potassium chloride saturated with silver chloride aqueous solution 3 mol/l (3 N) in aqueous solution for filling electrodes, Supelco®
Catalog # 1.04818.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Boiling Point:100 °C (1013 hPa)
- Density:1.13 g/cm3 (20 °C)
Specification Test Results
About this item
:
:74.55 g/mol
EINECS:231-211-8
UN:3082
:GHS09
Storage Temperature:Ambient
CAS Number:7447-40-7
Formula:KCl+AgCl
MDL Number:MFCD00011360
Density:1.13 g/cm³ (20 °C)
ADR:9 III
:M-Clarity™ quality level = MQ100