Order Entry
Switzerland
ContactUsLinkComponent
Tellurium standard solution, 10 mg/L Te in 5% nitric acid (from Te), standard for ICP, Specpure®
Tellurium standard solution, 10 mg/L Te in 5% nitric acid (from Te), standard for ICP, Specpure®
Catalog # 45265.AE
CAS Number:  
Tellurium standard solution, 10 mg/L Te in 5% nitric acid (from Te), standard for ICP, Specpure®
Catalog # 45265.AE
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    5% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Tellurium
  • Symbol:
    Te

Specifications

About this item

Certifications: Traceable to NIST.