Order Entry
Switzerland
ContactUsLinkComponent
 
VWR® ARISTAR®, Selenium Standard Solution, 10000 mg/L Se in dil. Nitric Acid (from Se), Standard for ICP
  455992B
 :  
undefined
VWR® ARISTAR®, Selenium Standard Solution, 10000 mg/L Se in dil. Nitric Acid (from Se), Standard for ICP
  455992B
 :  

 

  • Concentration:
    10000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Selenium
  • Symbol:
    Se

 

 

Se in HNO₃ 2 to 5%.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.