Order Entry
Switzerland
ContactUsLinkComponent
 
VWR®, Praseodymium standard solution, 10000 mg/L Pr in dil. nitric acid (from Pr6O11) ARISTAR® standard for ICP
Catalog # 455852N
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Praseodymium standard solution, 10000 mg/L Pr in dil. nitric acid (from Pr6O11) ARISTAR® standard for ICP
Catalog # 455852N
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    10000mg/L
  • Matrix:
    2-5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Praseodymium
  • Symbol:
    Pr

Specifications

About this item

Pr₆O₁₁ in HNO₃ 2 to 5%.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.