Order Entry
Switzerland
ContactUsLinkComponent
VWR® ARISTAR®, Niobium Standard Solution, 10000 mg/L Nb in Water with Hydrofluoric Acid (max. 1%) (from Nb₂O₅), Standard for ICP
VWR® ARISTAR®, Niobium Standard Solution, 10000 mg/L Nb in Water with Hydrofluoric Acid (max. 1%) (from Nb₂O₅), Standard for ICP
Catalog # 455752K
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Niobium Standard Solution, 10000 mg/L Nb in Water with Hydrofluoric Acid (max. 1%) (from Nb₂O₅), Standard for ICP
Catalog # 455752K
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    10000mg/L
  • Matrix:
    H₂O/tr HF
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Environmentally Preferable:
  • Element:
    Niobium
  • Symbol:
    Nb

Specifications

About this item

Nb₂O₅ in H₂O tr. HF.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.