Order Entry
Switzerland
ContactUsLinkComponent
 
VWR® ARISTAR®, Lithium Standard Solution, 200 mg/L Li in dil. Nitric Acid (from Li), Standard for Ion Chromatography
  458322E
 :  
undefined
VWR® ARISTAR®, Lithium Standard Solution, 200 mg/L Li in dil. Nitric Acid (from Li), Standard for Ion Chromatography
  458322E
 :  

 

  • Concentration:
    200 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Lithium
  • Symbol:
    Li

 

 

Li in H₂O tr.HNO₃.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.