Order Entry
Switzerland
ContactUsLinkComponent
ARISTAR®VWR® , Germanium Standard Solution, 10000 mg/L Ge in Water with hydrofluoric Acid (max. 1%) (from (NH4)2GeF6), Standard for ICP
ARISTAR®VWR® , Germanium Standard Solution, 10000 mg/L Ge in Water with hydrofluoric Acid (max. 1%) (from (NH4)2GeF6), Standard for ICP
Catalog # 455412Q
Supplier:  VWR Chemicals
CAS Number:  
undefined
ARISTAR®VWR® , Germanium Standard Solution, 10000 mg/L Ge in Water with hydrofluoric Acid (max. 1%) (from (NH4)2GeF6), Standard for ICP
Catalog # 455412Q
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    H₂O/tr HF
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Environmentally Preferable:
  • Element:
    Germanium
  • Symbol:
    Ge

Specifications

About this item

(NH₄)₂GeF₆ in H₂O tr. HF.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.