VWR® ARISTAR®, Beryllium Standard Solution, 1000 mg/L Be in dil. Nitric Acid, Standard for ICP
Nome del fornitore: VWR Chemicals
Certificati
Informazioni su questo articolo
BeO.(Be(OAc)₂)₃) in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.
Informazioni correlate
Certificati
Cerca per numero di lotto125 ml Plastic bottle for liquids DIN 24 (ZPACK15150125) 500 ml Plastic bottle for liquids DIN 28 (ZPACK15150500)
Packaging Information
VWR® for Calibrations
Product Literature
Frequently Bought Together