

Titanium(III) chloride 15% in hydrochloric acid 10% for synthesis, Sigma-Aldrich®
: Merck
:7705-07-9
:
:Cl₃Ti
:MFCD00011266
:154.24 g/mol
:1.21 g/cm³ (20 °C)
:231-728-9
:1789
:8,II
:M-Clarity™ quality level = MQ200
:GHS05
:GHS07
- Assay (cerimetric TiCl₃):14.5 - 15.5 %
- Density:1.21 g/cm3 (20 °C)
- pH:- 1 (H2O, 20 °C) (undiluted)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Frequently Bought Together