Tetrapropylammonium hydroxide 40% in aqueous solution for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:4499-86-9
:
Formula:C₁₂H₂₉NO
MDL Number:MFCD00009360
:203.37 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:224-800-6
UN:3267
:GHS05
Storage Temperature:Refrigerator
Specification Test Results
- Solubility:(20 °C) soluble
- Boiling Point:102 °C (1013 hPa)
- Density:1.00 g/cm3 (20 °C)
- pH:7 (H2O, 20 °C) (undiluted)
Specification Test Results
Frequently Bought Together