Rifampicin, orange powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:13292-46-1
:
Formula:C₄₃H₅₈N₄O₁₂
MDL Number:MFCD00151389
:822.95 g/mol
Merck Index:12,08382
Melting Pt:183…188 °C (dec., lit.)
EINECS:236-312-0
:GHS07
Specification Test Results
- Synonyms:Rifamycin AMP; Rifampicin; 3-(4-Methylpiperazinyliminomethyl)rifamycin SV; Rifampicin
- Form:Orange Powder
- Storage:Store at +4 °C
- Isoelectric Point:4,8 (in water) (Lit.)
- Loss on Drying:≤2%
- Optical Rotation:[α]D25°=+10,6° (c=0,5% in CDCl3)(Lit.)
Specification Test Results
Frequently Bought Together