
G418 disulphate for biochemistry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108321-42-2
:
Formula:C₂₀H₄₄N₄O₁₈S₂
MDL Number:MFCD00058314
:692.72 g/mol
:GHS08
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Water:≤10 %
- Potency:≥650 µg/mg
Specification Test Results
Frequently Bought Together