Order Entry
Switzerland
ContactUsLinkComponent
 
Boron trifluoride diethyl ether complex BF₃ ∼ 48%, AcroSeal®
 
undefined
Boron trifluoride diethyl ether complex BF₃ ∼ 48%, AcroSeal®
 

 

  • Appearance:
    Clear colorless to straw yellow Liquid
  • Refractive index:
    1.3429 to 1.3449 (20°C, 589 nm)
  • Assay:
    47.5 to 49 % (BF3)
  • GC:
    ≥98.0 %

 

Frequently Bought Together