Order Entry
Switzerland
ContactUsLinkComponent
Lactose monohydrate, AnalaR NORMAPUR® analytical reagent
Catalog # 101394S
Supplier:  VWR Chemicals
CAS Number:  
undefined
Lactose monohydrate, AnalaR NORMAPUR® analytical reagent
Catalog # 101394S
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Specific optical rotation (10 %; water):
    52.0 - 52.8 °
  • Acidity:
    Max. 0.005 meq/g
  • Ignition residue (SO4):
    Max. 0.1 %
  • Insolubility in water:
    Max. 50 ppm
  • Water:
    4.8 - 5.4 %
  • As (Arsenic):
    Max. 1 ppm
  • Cu (Copper):
    Max. 0.5 ppm
  • Fe (Iron):
    Max. 1 ppm
  • Pb (Lead):
    Max. 0.5 ppm

Specification Test Results

About this item

 :
 :360.32 g/mol
Melting Pt:223 °C
Storage Temperature:Ambient
EINECS:200-559-2
CAS Number:10039-26-6
Formula:C₁₂H₂₂O₁₁·1H₂O