Order Entry
Switzerland
ContactUsLinkComponent
Isohexane contains <5% n-Hexane for pesticide residue analysis
Irritant
Environment
Flammable
Health
Isohexane contains <5% n-Hexane for pesticide residue analysis
Certificates
undefined
Irritant
Environment
Flammable
Health
Isohexane contains <5% n-Hexane for pesticide residue analysis

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.01 % (Coulometric)
  • GC:
    (ECD) Passes test, (NPD) Passes test, (FID) Passes test
  • Residue after evaporation:
    ≤2 ppm

Specification Test Results

Related Information

Frequently Bought Together