



Solvent ASTM D2896, Chlorobenzene/Acetic acid-mixture (2/1 v/v), TBN solvent, VOLUSOL®
Catalog # 799306A
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay (Acetic acid) (V/V):33.08 - 33.58 %(V)
- Assay (Chlorobenzene) (V/V):66.41 - 66.91 %(V)
Specification Test Results
About this item
:
Boiling Pt:>118 °C (1013 hPa)
Density:1.08 g/cm³ (20 °C)
Flash Pt:32 °C (closed cup)
Storage Temperature:Ambient
UN:2924
ADR:8,II
:GHS02
:GHS05
:GHS07
:GHS09