Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Picric acid
Picric acid 1.2% in aqueous solution, TECHNICAL
Picric acid 1,2% in aqueous solution, TECHNICAL
84512.260
:
VWR Chemicals
:
Picric acid 1,2% in aqueous solution, TECHNICAL
84512.260
:
VWR Chemicals
:
Pack type:
Plastic bottle
Pk:
500 mL
Seq:
2725
Assay:
1.0 - 1.2%
:
:
229.11 g/mol
:
201-865-9
:
88-89-1
:
C₆H₃N₃O₇
:
MFCD00007102