

Ethanol denatured 70% ≥70% (v/v), TechniSolv®
83805.440
:
- Pack type:Plastic drum
- Pk:20 L
- Seq:5302
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
- Assay:Min. 67 %
- Density (20/20):0.884 - 0.89 g/cm³
:
:46.07 g/mol
:Ambient
:200-578-6
:1170
:3,II
:GHS02
:GHS07
:64-17-5
:H₃CCH₂OH
:MFCD00003568