
Methyl red 0.2 g/l in ethanol, TECHNICAL
Catalog # 34302.231
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- Seq:2590
Specifications
Specification Test Results
- Identification:Passes test
Specification Test Results
About this item
:
Flash Pt:20 °C (closed cup)
Storage Temperature:Ambient
EINECS:207-776-1
ADR:3,II
:269.3 g/mol
UN:1170
:GHS02
CAS Number:493-52-7
Formula:C₁₅H₁₅N₃O₂
MDL Number:MFCD00002425