Bromocresol green 0,04% in ethanol, TECHNICAL
Glass bottle, 250 mL
Supplier: VWR Chemicals
Bromocresol green 0,04% in ethanol, TECHNICAL
Catalog # 34301.237
Supplier: VWR Chemicals
CAS Number: 76-60-8
Jump to:
Product Details & Documents
- Transition range: pH 3.8-5.4
Synonyms: 3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- CAS Number:76-60-8
- Formula:C₂₁H₁₄Br₄O₅S
- MDL Number:MFCD00005874
- Molecular Weight:698.02 g/mol
- Storage Temperature:Ambient
- EINECS:200-972-8
- UN:1170
- ADR:3,II
Documents
Certificates
Search by Lot Number250 ml Glass bottle for liquids DIN 32 (ZPACK13415030)
Packaging Information
P20_VWRC34301.237_SPEC_EN.pdf spec_VWRC34301.237 spec_VWRC34301.237
Product specification
VWR® Indicators
Product Literature
Specifications
Specification Test Results
- Identification:Passes test
Specifications
- Pack type:Glass bottle
- Pk:250 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



