Order Entry
Switzerland
ContactUsLinkComponent
Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) in toluene 90% (v/v)/methanol 10% (v/v), AVS TITRINORM volumetric solution
Flammable
Health
Irritant
Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) in toluene 90% (v/v)/methanol 10% (v/v), AVS TITRINORM volumetric solution
Catalog # 28532.267
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Health
Irritant
Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) in toluene 90% (v/v)/methanol 10% (v/v), AVS TITRINORM volumetric solution
Catalog # 28532.267
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.0998 - 0.1002 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :259.48 g/mol
Storage Temperature:Refrigerator
EINECS:218-147-6
UN:1992
ADR:3 (6.1),II
 :GHS02
 :GHS08
 :GHS07
CAS Number:2052-49-5
Formula:C₁₆H₃₇NO
MDL Number:MFCD00009425