Order Entry
Switzerland
ContactUsLinkComponent
 
Saponin, GPR RECTAPUR®
Catalog # 27534.187
Supplier:  VWR Chemicals
CAS Number:  
undefined
Saponin, GPR RECTAPUR®
Catalog # 27534.187
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Foam test (5 %; water):
    2 - 6 cm
  • Ignition residue (SO4):
    Max. 10 %
  • Insolubility in water:
    Max. 0.5 %

Specification Test Results

About this item

 :
Density:1.015…1.02 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-462-6
CAS Number:8047-15-2
MDL Number:MFCD00081981