

Methyl methacrylate ≥99.5% stabilised, GPR RECTAPUR®
Catalog # 25600.296
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Assay:Min. 99.5 %
- Acidity:Max. 0.0004 meq/g
- Colouration:Max. 5 APHA
- Density (20/4):0.942 --> 0.944
- Stabilizer (MEHQ):10 --> 25 ppm
- Water:Max. 0.05 %
Specification Test Results
About this item
:
:100.12 g/mol
Merck Index:12,06005
Boiling Pt:100 °C (1013 hPa)
Melting Pt:–48 °C
Density:0.93 g/cm³ (20 °C)
Flash Pt:8 °C
Storage Temperature:Ambient
EINECS:201-297-1
UN:1247
ADR:3,II
:Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
REACH:01-2119452498-28
:GHS02
:GHS07
CAS Number:80-62-6
Formula:CH₂=C(CH₃)COOCH₃
MDL Number:MFCD00008587