

Chromicsulphuric acid mixture (0.8% chromium(VI) oxide in sulphuric acid 95%), TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1333-82-0
:
Formula:CrO₃
:99.99 g/mol
EINECS:215-607-8
UN:3264
ADR:8,II
:GHS05
:GHS08
Specification Test Results
- Identification:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle Safebreak for liquids DIN 45 (ZPACK13415111) 2,5L Glass bottle Safebreak for liquids DIN 45 (ZPACK13415250)
Packaging Information
28576995.pdf spec_VWRC25333.247 spec_VWRC25333.247 spec_VWRC25333.327
Product specification
Frequently Bought Together