
Diethylene glycol monobutyl ether ≥98%, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:112-34-5
:
Formula:CH₃(CH₂)₃OCH₂CH₂OCH₂CH₂OH
MDL Number:MFCD00002881
:162.23 g/mol
Merck Index:13,01556
Boiling Pt:230.4 °C (1013 hPa)
Melting Pt:–68 °C
Density:0.954 g/cm³ (20 °C)
Flash Pt:100 °C
EINECS:203-961-6
:GHS07
Storage Temperature:Ambient
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Assay:Min. 98 %
- Refractive index (20°C):1.431 - 1.433
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101)
Packaging Information
spec_VWRC23831.291 spec_VWRC23831.291 spec_VWRC23831.460
Product specification
Frequently Bought Together