ContactUsLinkComponent
Apiezon® sealing compound Q
Apiezon® sealing compound Q
Catalog # 22844.298
Supplier:  M I MATERIALS
Apiezon® sealing compound Q
Catalog # 22844.298
Supplier:  M I MATERIALS
Specifications
  • Description:
    Sealing Compound Q
  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
Specifications
About this item
Used to seal joints and gaps and to effect temporary repairs. Good for sealing flat unground joints. Can be moulded by hand as it is a putty-like substance. For use at ambient temperatures.
  • Vapour pressure 1 x 10-4 torr at 20°C