ContactUsLinkComponent
VWR® ARISTAR® Multi-Component Anion Mix 9, Avantor
VWR® ARISTAR® Multi-Component Anion Mix 9, Avantor
Catalog # 458164C
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR® Multi-Component Anion Mix 9, Avantor
Catalog # 458164C
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pack type:
    Plastic bottle
  • Pk:
    1 SET
  • Description:
    Multi-Component Anion Mix 9
Specifications
About this item
Two solutions - a nitrite standard solution and a 6 component multi-element standard solution.
Br 40 ppm   NO3 40 ppm
Cl 40 ppm   PO4 40 ppm
F 20 ppm   SO4 40 ppm
NO2 40 ppm
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory
Supplied with certificate of analysis.