Order Entry
Switzerland
ContactUsLinkComponent
VWR® ARISTAR®, Interference Check B Nitric Acid 2% for ICP-MS
VWR® ARISTAR®, Interference Check B Nitric Acid 2% for ICP-MS
Catalog # 456562K
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Interference Check B Nitric Acid 2% for ICP-MS
Catalog # 456562K
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Interference check B in nitric acid 2% ARISTAR® for ICP-MS

Specifications

About this item

11 Elements in 2% nitric acid solution.

11 Elements : 10 mg/L As, Cd, Se Zn; 20 mg/L Ag, C, Cr, Cu, Mn, Ni, V

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery: Supplied with certificate of analysis.