Order Entry
Switzerland
ContactUsLinkComponent
VWR® ARISTAR®, Interference Check A in Nitric Acid 1% for ICP-MS
VWR® ARISTAR®, Interference Check A in Nitric Acid 1% for ICP-MS
Catalog # 456552Y
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Interference Check A in Nitric Acid 1% for ICP-MS
Catalog # 456552Y
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Interference check A in nitric acid 1% ARISTAR® for ICP-MS

Specifications

About this item

(12 Elements nitric acid 1% solution)

12 Elements:18000 mg/L Cl; 1000 mg/L Al, K, Mg, P, S; 2000 mg/L C; 3000 mg/L Ca; 2500 mg/L Fe, Na; 20 mg/L Mo, Ti

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery: Supplied with certificate of analysis.